Learn More
Selleck Chemical LLC Ginkgolic Acid-1mg
SDP
Supplier:  Selleck Chemical LLC S94321MG
Encompass_Preferred
Ginkgolic acid, toxic phenolic compounds present in the fruits and leaves of Ginkgo biloba L., is a potent sumoylation inhibitor also reported to inhibit histone acetylation transferase (HAT).
Catalog No. 50-238-9930