Learn More
Medchemexpress LLC Ginkgolic acid C17:1 | 111047-30-4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN21165MG
Encompass_Preferred
Ginkgolic acid C17:1 is a fatty acid synthase (FAS) inhibitor with an IC50 of 10.5 μM. It exhibits anti-tumor activity by inhibiting the phosphorylation of STAT3 and inducing apoptosis. This compound also blocks the interaction between S-RBD and ACE2, showing anti-SARS-CoV-2-S pseudovirus activity. Additionally, it inhibits the biofilm formation of enterohemorrhagic Escherichia coli and Staphylococcus aureus.
  • Inhibits fatty acid synthase (FAS).
  • Exhibits anti-tumor activity.
  • Induces apoptosis.
  • Blocks S-RBD and ACE2 interaction.
  • Shows anti-SARS-CoV-2-S pseudovirus activity.
  • Inhibits biofilm formation of enterohemorrhagic Escherichia coli.
  • Inhibits biofilm formation of Staphylococcus aureus.
Catalog No. 50-004-21684