Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginkgolic acid C17:1 | 111047-30-4 | 99.8% | 10 MG
SDP

Supplier:  Medchemexpress LLC HYN211610MG

Encompass_Preferred

Ginkgolic acid C17:1 is a naturally derived alkyl phenolic compound supplied for research use with reported activity against STAT3 signaling and inhibitory effects on viral spike-ACE2 interactions. It is provided as a high-purity research standard for biochemical assays and mechanism-of-action studies.

  • natural alkyl phenolic acid useful for biochemical research.
  • reported antitumor and antiviral activity in literature.
  • high purity (LCMS) suitable for analytical and biological testing.
  • molecular weight 374.56 g/mol for stoichiometric calculations.
  • store at -20°C and protect from light to preserve stability.

Catalog No. 50-002-96620


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.