Learn More
Cayman Chemical GInkgolIde C 10mg
SDP
Supplier:  Cayman Chemical 1987210 MG
Encompass_Preferred
A selective PAF receptor antagonist (IC50 = 37.6 μM); increases lipolysis and inhibits adipogenesis in adipocytes via the activated AMPK pathway
Catalog No. 50-298-3078