Learn More
Cayman Chemical GInkgolIde J 10mg
SDP
Supplier:  Cayman Chemical 2916310 MG
Encompass_Preferred
A terpene trilactone with diverse biological activities; inhibits PAF-induced platelet in rabbit and human platelet-rich plasma (IC50s = 27 and 43.5 μg/ml, respectively); inhibits Aβ42-induced cell death in primary rat hippocampal neurons and Aβ42-induced impairment of LTP in the mouse CA1 region at 1 μM
Catalog No. 50-294-1946