Learn More
Apexbio Technology LLC Ginsenoside F1 53963-43-2 20mg
SDP
Supplier:  Apexbio Technology LLC N156920
Encompass_Preferred
Ginsenoside F1 (CAS 53963-43-2) is a saponin compound isolated from Panax ginseng C A Mey This molecule is known to modulate cellular signaling pathways associated with neuroprotection anti-inflammatory responses and regulation of oxidative stress Studies indicate that Ginsenoside F1 influences pathways such as PI3K/Akt and MAPK contributing to its effects on cellular proliferation and apoptosis The compound is widely utilized in biomedical research focused on neurodegenerative disorders inflammation and metabolic diseases Ginsenoside F1 is soluble in water methanol and ethanol and should be stored in a cool dry sealed environment
Catalog No. 50-000-91660