Learn More
Medchemexpress LLC HY-N0598 5mg Medchemexpress, Ginsenoside F1 CAS:53963-43-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN05985MG
Encompass_Preferred
Medchemexpress, HY-N0598 5mg Ginsenoside F1 CAS:53963-43-2 Ginsenoside F1, an enzymatically modified derivative of Ginsenoside Rg1, demonstrates competitive inhibition of CYP3A4 activity and weaker inhibition of CYP2D6 activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3632