Learn More
Selleck Chemical LLC Ginsenoside F1 S9299-1MG
SDP
Supplier:  Selleck Chemical LLC S92991MG
Encompass_Preferred
Ginsenoside F1 a pharmaceutical component of ginseng is known to have antiaging antioxidant anticancer and keratinocyte protective effects Ginsenoside F1 exhibits competitive inhibition of the activity of CYP3A4 with Ki values of 67 8 16 2 M Ginsenoside F1 also exhibited a weaker inhibition of the activity of CYP2D6
Catalog No. 50-000-23678