Learn More
Medchemexpress LLC HY-125848 5mg Medchemexpress, Ginsenoside F2 CAS:62025-49-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1258485MG
Encompass_Preferred
Medchemexpress, HY-125848 5mg Ginsenoside F2 CAS:62025-49-4 Ginsenoside F2, a metabolite from Ginsenoside Rb1, induces apoptosis accompanied by protective autophagy in breast cancer stem cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9705