Learn More
Medchemexpress LLC Ginsenoside F3 | 62025-50-7 | 770.99 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN060010MM/1ML
Encompass_Preferred
Ginsenoside F3, a component of PPTGs (a minor saponin in the leaves of Panax ginseng), has immunoenhancing activity by regulating the production and gene expression of type 1 cytokines (IL-2, IFN-gamma) and type 2 cytokines (IL-4 and IL-10).
  • Purity: 99.84%.
Catalog No. 50-003-89034