Learn More
Medchemexpress LLC Ginsenoside F5 | 189513-26-6 | MFCD22124710 | 99.9% | 770.99 g·mol⁻¹ | C41H70O13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1082771MG
Encompass_Preferred
Ginsenoside F5 is a natural triterpenoid saponin isolated from the flower buds of Panax ginseng. It has demonstrated antiproliferative activity against HL-60 leukemia cells by inducing apoptosis and is used as a research reagent in biochemical and pharmacological studies of ginsenoside activity and anticancer mechanisms.
  • High purity: 99.9%.
  • Molecular formula C41H70O13 and molecular weight 770.99 g·mol⁻¹.
  • Suitable for in vitro biochemical and cell-based assays.
  • Supplied in small, precisely measured quantities for analytical and pharmacological research.
  • Supports mechanistic studies of apoptosis and anticancer pathways.
Catalog No. 50-002-42682