Learn More
Cayman Chemical GInsenosIde Rc 10mg
SDP
Supplier:  Cayman Chemical 2908810 MG
Encompass_Preferred
A saponin; prevents UVB-induced increases in ROS levels in HaCaT keratinocytes by 34.3, 54.4, and 65.4% when used at concentrations of 5, 12, and 30 µM, respectively; reduces increases in LPS-induced expression of Tnf, Il1b, and Ifnb1 in RAW 264.7 cells at 40 UG/ml; reduces gastric ulcer and joint lesion indices in mouse models of HCl/ethanol-induced gastritis and collagen-induced arthritis, respectively, as well as reduced increased serum levels of AST induced by LPS/D-galactosamine in a mouse model of hepatitis, at 20 mg/kg; increases GABA-induced inward current in Xenopus oocytes expressing recombinant human α1β1ɣ2S subunit-containing GABAA receptors (EC50 = 53.2 µM)
Catalog No. 50-294-0425