Learn More
Apexbio Technology LLC Ginsenoside Rd 52705-93-8 20mg
SDP
Supplier:  Apexbio Technology LLC N161620
Encompass_Preferred
Ginsenoside Rd (CAS 52705-93-8) is a protopanaxadiol-type saponin derived from Panax ginseng This small molecule has been reported to modulate multiple signaling pathways associated with cell proliferation apoptosis and inflammation Experimental evidence suggests that Ginsenoside Rd can suppress tumor cell growth possibly by interfering with cell cycle progression and promoting apoptotic mechanisms Due to these properties it is utilized in biomedical research to investigate mechanisms underlying cancer development and to explore novel therapeutic approaches targeting tumor progression
Catalog No. 50-000-90708