Learn More
Cayman Chemical GInsenosIde Rd 5mg
SDP
Supplier:  Cayman Chemical 153295 MG
Encompass_Preferred
A protopanaxadiol that has diverse in vitro and in vivo effects, including cardioprotective, neuroprotective, and anti-inflammatory actions
Catalog No. 50-297-0183