Learn More
Apexbio Technology LLC Ginsenoside Re 52286-59-6 20mg
SDP
Supplier:  Apexbio Technology LLC N161520
Encompass_Preferred
Ginsenoside Re (CAS 52286-59-6) is a natural saponin isolated from Panax notoginseng In neurobiological research it has been shown to reduce -amyloid (A ) levels and inhibit the c-Jun N-terminal kinase (JNK) signaling pathway as well as the activity of the NF- B transcription factor These properties implicate Ginsenoside Re in the modulation of neuroinflammatory and neurodegenerative processes The compound is commonly utilized in experimental models to investigate the molecular mechanisms underlying Alzheimer s disease neurodegeneration and inflammatory signaling pathways
Catalog No. 50-000-90700