Learn More
Medchemexpress LLC HY-N0601 5mg Medchemexpress, Ginsenoside Rf CAS:52286-58-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN06015MG
Encompass_Preferred
Medchemexpress, HY-N0601 5mg Ginsenoside Rf CAS:52286-58-5 Ginsenoside Rf is a trace component of ginseng root. Ginsenoside Rf inhibits N-type Ca 2+ channel . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3633