Learn More
AdipoGen Ginsenoside Rg3 (5 mg)
SDP
Supplier:  AdipoGen CDXG0220M005
Encompass_Preferred
Chemical. CAS: 14197-60-5. Formula: C42H72O13. MW: 785.01. Ginsenoside Rg3 is a panaxadiol found in white and red P. ginseng. Show a broad range of biological in vitro and in vivo effects, including anticancer, antidiabetic, neuroprotective, antioxidant, anti-hypertensive, and anti-inflammatory actions. The anticancer mechanisms include induction of apoptosis and autophagy, inhibition of proliferation, inhibition of metastasis and angiogenesis, cell cycle arrest, immunomodulatory effects, sensitization to radiation, reducing multidrug resistance and inducing genotoxicity to the cancer cells. Ginsenoside Rg3 has been shown to inhibit the 5-HT3A and alpha3beta4 nACh receptors, the voltage-dependent Ca2+, K+, and Na+ channel currents. It is a scavenger of hydroxyl radicals and downregulated the expression of DNA methyltransferases, reducing global DNA methylation, modifying the methylation of the promoter region of some relevant genes in cancer.
Catalog No. 50-205-4482