Learn More
Selleck Chemical LLC Ginsenoside Rg5 E2459-1MG
SDP
Supplier:  Selleck Chemical LLC E24591MG
Encompass_Preferred
Ginsenoside Rg5 the main component of Red ginseng blocks binding of IGF-1 to its receptor with an IC50 of 90 nM Ginsenoside Rg5 also inhibits the mRNA expression of COX-2 via suppression of the DNA binding activities of NF- B p65
Catalog No. 50-000-40142