Learn More
Apexbio Technology LLC Ginsenoside Rh1 63223-86-9 20mg
SDP
Supplier:  Apexbio Technology LLC N161020
Encompass_Preferred
Ginsenoside Rh1 (CAS 63223-86-9) is a small-molecule modulator targeting nuclear receptor pathways such as peroxisome proliferator-activated receptor gamma (PPAR- ) It is designed to modulate these receptors thereby regulating inflammatory signaling cascades Ginsenoside Rh1 exerts its biological activity primarily through inhibition of inflammatory cytokine and mediator expression via the suppression of inflammation-associated signaling pathways Based on these pharmacological properties Ginsenoside Rh1 holds research potential in the regulation of inflammation metabolic pathway investigation and nuclear receptor pathway modulation
Catalog No. 50-000-91662