Learn More
Medchemexpress LLC Ginsenoside Rh2 | 78214-33-2 | 622.87 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYN0605
Encompass_Preferred
Ginsenoside Rh2 is a promising candidate for anti-tumor drug development, promoting cancer cell apoptosis through multiple pathways by activating caspase-8 and caspase-9. It triggers p53-dependent Fas expression and subsequent activation of caspase-8, as well as a p53-independent caspase-9-mediated intrinsic pathway to cause cancer cell death.
  • Induces activation of caspase-8 and caspase-9.
  • Promotes cancer cell apoptosis in a multi-path manner.
  • Triggers p53-dependent Fas expression.
  • Activates p53-independent caspase-9-mediated intrinsic pathway.
  • Exhibits cytotoxic activity in various human tumor cell lines.
  • Shown to reduce tumor sizes and increase survival in animal models.
  • Purity of 99.28%.
  • Appears as a white to off-white solid.
  • Derived from natural sources (Panax ginseng C. A. Meyer).
Catalog No. 50-003-90217