Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginsenoside Rk2 | 364779-14-6 | 1 MG
SDP

Supplier:  Medchemexpress LLC HYN25041MG

Encompass_Preferred

Ginsenoside Rk2 is a dammarane glycoside isolated from processed ginseng. Intended for research use only, it has 99.25% purity and appears as an off-white to light yellow solid.

  • Molecular weight: 604.86
  • Formula: C36H60O7
  • Source: Panax ginseng C. A. Meyer (Araliaceae plants)
  • Storage: -20°C, protected from light. In solvent, -80°C for 6 months or -20°C for 1 month.
  • In vitro solubility: Soluble in DMSO (50 mg/mL, 82.66 mM), requires sonication.
  • In vivo solubility: Protocols for suspended (1.25 mg/mL in DMSO/PEG300/Tween-80/Saline) or clear solution (≥ 1.25 mg/mL in DMSO/Corn Oil).
  • Cellular effect: IC50 > 10 μM against human A549, HL-60, MCF7, and SW480 cells.
  • Classifications: Natural products, terpenoids, triterpenes, inflammation/immunology.

Catalog No. 50-004-20084


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.