Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ginsenoside Rk2 | 364779-14-6 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYN25045MG

Encompass_Preferred

Ginsenoside Rk2 is an off-white to light yellow solid with 99.25% purity. It is a dammarane glycoside isolated from processed ginseng (Sun Ginseng) and categorized under Terpenoids, specifically Triterpenes, with an initial source from the *Panax ginseng C. A. Meyer* plant. Its molecular weight is 604.86 and chemical formula is C36H60O7.

  • Purity of 99.25%.
  • Molecular weight of 604.86.
  • Chemical formula is C36H60O7.
  • Isolated from processed ginseng.
  • Classified as a dammarane glycoside, terpenoid, and triterpene.
  • Shows IC50 > 10 μM for cytotoxicity against human A549, HL-60, MCF7, and SW480 cells.
  • Soluble in DMSO (50 mg/mL, with ultrasonic aid).
  • Provides in vivo dissolution protocols.

Catalog No. 50-004-21440


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.