Learn More
Medchemexpress LLC HY-N0906 25mg Medchemexpress, Ginsenoside Rk3 CAS:364779-15-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN090625MG
Encompass_Preferred
Medchemexpress, HY-N0906 25mg Ginsenoside Rk3 CAS:364779-15-7 Ginsenoside Rk3 is present in the roots Panax notoginseng herbs. Ginsenoside Rk3 significantly inhibits TNF-α-induced NF-κB transcriptional activity, with an IC50 of 14.24±1.30 μM in HepG2 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2827