Learn More
Cayman Chemical GIrard Reagent P 5g
SDP
Supplier:  Cayman Chemical 6015415 G
Encompass_Preferred
A cationic hydrazine reagent; has been used as a derivatization reagent to quantify glycans, oxysterols, and 5-methylcytosine derivatives
Catalog No. 50-289-6715