Learn More
ABclonal Technology GIRK2KCNJ6 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2439120UL
Encompass_Preferred
This gene encodes a member of the G protein-coupled inwardly-rectifying potassium channel family of inward rectifier potassium channels This type of potassium channel allows a greater flow of potassium into the cell than out of it These proteins modulate many physiological processes including heart rate in cardiac cells and circuit activity in neuronal cells through G-protein coupled receptor stimulation Mutations in this gene are associated with Keppen-Lubinsky Syndrome a rare condition characterized by severe developmental delay facial dysmorphism and intellectual disability
Catalog No. 50-000-10602