Learn More
Medchemexpress LLC Gisadenafil besylate | 334827-98-4 | 677.79 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10861910MG
Encompass_Preferred
Gisadenafil besylate (UK 369003-26) is a specific, orally active phosphodiesterase 5 (PDE5) inhibitor with an IC50 of 3.6 nM and prevents degradation of cyclic guanosine monophosphate (cGMP).
  • Specific, orally active phosphodiesterase 5 (PDE5) inhibitor.
  • IC50 for PDE5A is 3.6 nM.
  • Inhibits PDE1A with an IC50 of 9.1 μM, showing an approximately 2500-fold difference in specificity for PDE5A.
  • In vivo, Gisadenafil (2 mg/kg; intraperitoneal injection) largely restores the normal increase in cortical flow following hypercapnia in Tat-tg mice (17.5% above baseline).
  • Restores the dilation of small (<25 μm) arterioles following hypercapnia.
Catalog No. 50-003-44305