Learn More
Medchemexpress LLC Gitogenin | 511-96-6 | MFCD00273328 | 99.8% | 432.64 | C27H44O4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN257410MG
Encompass_Preferred
Gitogenin is a natural steroid derived from Tribulus longipetalus that is used as a biochemical inhibitor in enzyme and metabolic studies. It selectively inhibits UDP-glucuronosyltransferase 1A4 (UGT1A4) and α-glucosidase and is supplied as a high-purity analytical material for laboratory research.
  • Natural steroid isolated from Tribulus longipetalus.
  • Selective inhibitor of UDP-glucuronosyltransferase 1A4 (UGT1A4) and α-glucosidase.
  • Reported IC50 values: 0.69 μM for UGT1A4, 37.2 μM for α-glucosidase.
  • High purity suitable for research and analytical applications.
  • Supplied in small quantities appropriate for analytical assays.
Catalog No. 50-002-89626