Learn More
AdipoGen Gitoxin (5 mg)
SDP
Supplier:  AdipoGen AGCN20485M005
Encompass_Preferred
Chemical. CAS: 4562-36-1. Formula: C41H64O14. MW: 780.9. Isolated from Digitalis purpurea L. Cardiac glycoside. Natural metabolite of digitoxin. Starting material for the synthesis of gitoxin derivatives with activity as cardiac glycosides. Uncompetitive inhibitor of the Na+/K+ ATPase activity. Inhibits growth of selected human cancer cells.
Catalog No. 50-205-3668