Learn More
Cayman Chemical GITR EXrAClular DomaInhu 100ug
SDP
Supplier:  Cayman Chemical 32017100 UG
Encompass_Preferred
Source: Active recombinant human C-terminal His-tagged GITR expressed in HEK293 cells • Amino acids: 26-161 • MW: 16 kDa
Catalog No. 50-294-9091