Learn More
Medchemexpress LLC Givinostat (ITF-2357) | 497833-27-9 | 98.7% | 421.49 g·mol⁻¹ | C24H27N3O4 | 10 MG

Supplier: Medchemexpress LLC HY1484210MG
Givinostat (ITF-2357) is a histone deacetylase (HDAC) inhibitor used in preclinical research. It selectively inhibits class I HDAC enzymes and is applied in studies investigating inflammatory and muscular disorders, including Duchenne muscular dystrophy (DMD).
- Inhibits HDAC1 (IC50 198 nM) and HDAC3 (IC50 157 nM).
- High purity (98.7%) suitable for research use.
- Molecular weight 421.49 g·mol⁻¹.
- Chemical formula C24H27N3O4.
- Supplied as a 10 mg quantity for small-scale experiments.
- Applicable to in vitro and preclinical in vivo studies focused on epigenetic modulation.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.