Learn More
Cayman Chemical GJ-103sodIum salt 10mg
SDP
Supplier:  Cayman Chemical 2248210 MG
Encompass_Preferred
An inducer of stop codon read-through; restores the kinase activity of ATM in patient-derived A-T lymphoblastoid cell lines containing homozygous TAG, TGA, or TAA nonsense mutations in ATM at 10 and 30 μM
Catalog No. 50-296-4112