Learn More
Aviva Systems Biology Corp GJD3 Antibody - C-terminal region (ARP36632_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36632_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GJD3. It was validated on Western Blot. . Alternate names: CX31.9, Cx30.2, GJA11, GJC1ApplicationWB. Species reactivity: Human
Catalog No. 50-284-506