Learn More
ORS Nasco GL DRY BX HYPALON 15MIL 1PR
Supplier:  ORS Nasco 0688Y1532/9Q
Encompass_Preferred
Honeywell part# 8Y1532/9Q. Honeywell CSM Glove Box Gloves 8 in. Port Size: 9.75 Hand: Hand specific Thickness: 15 mil 0.38 mm White Latex Free: Yes Chlorosulfonated Polyethylene (CSM) Port Size: 20.3 cm 8 in. Certifications/Compliance: EN 388 EN 374 Circumference Hand: 9.75 in. 25 cm Exterior Finish: Smooth
Catalog No. 50-255-1653