Learn More
Bellco Glass, Inc. GL45 6 Baffle Shake Flask,1000mL With Membrane Screw Cap
SDP
Supplier:  Bellco Glass, Inc. 254501000
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. 3 Side baffles positioned at 120 degree and 3 bottom baffles offset from the side baffles. GL45 polypropylene membrane cap has a sealed in 0.2u PTFE membrane. Cap can be tightened during autoclaving.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassIncludes Polypropylene Membrane Cap And Pouring RingSize: 1LDimensions: 128x200mmCase Quantity: 6
Catalog No. 50-194-5544