Learn More
Selleck Chemical LLC Glafenine E4889-50MG
SDP
Supplier:  Selleck Chemical LLC E488950MG
Encompass_Preferred
Glafenine (Glafenin) is a non-steroidal anti-inflammatory drug (NSAID) and is a non-narcotic analgesic agent that not only acts as a proteostasis modulator but is also one of the most potent NSAID correctors of F508del-CFTR It exhibits COX-2 inhibition in the arachidonic acid pathway and rescues class 2 CFTR mutants
Catalog No. 50-000-41560