Learn More
Glas Col LLC Heating Mantle
SDP
Supplier:  Glas Col LLC 100A O1123
Encompass_Preferred
Silicone impregnated glass fabric heating mantle for 3L spherical flask, covers both the top and bottom of 1-2-3 neck flask, 200W top half, 500W bottom half. 115V, CSA. Top and bottom held together with zipper to prevent heat loss. The side split in the top allows removal without disturbing the flask.
Catalog No. NC1283375