Learn More
Ace Glass, Inc. Glass baffle, 10mm diax250mm length shaft, fits #11Ace-Thred adapter. Baffle dimensions: 18mmx60mmx6.4mm, 310mm overall length.
SDP
Supplier:  Ace Glass, Inc. 1217710
Encompass
18mm X 60mm X 6.4mm Glass Baffle. Overall length: 310mm. Use with #11 Ace-Thred adapters to mate glass or PTFE (stainless steel core) baffles to any standard taper joint. Baffles can be adjusted vertically, or rotated to increase agitation.
Catalog No. 50-977-539