Learn More
Bellco Glass, Inc. Glass Ball Spinner Only,250mL No Ctr Cap
SDP
Supplier:  Bellco Glass, Inc. 196594025
Encompass_Preferred
Description: Glass Ball Spinner Flask Only. MADE IN THE USASize: 250mLMaterial: Type 1 Class A Borosilicate GlassDimensions: 85mm OD x 140mm Overall Height, 164mm across sidearmsTwo 32mm Sidearms angled 55° offset center70mm Screw Thread Center NeckDimpled BottomGraduations: 50mL to 250mL, every 25mL – White Ink (Approx Volumes)Supply with: Two 32mm PP Blue Screw Caps & PP Blue Pouring Rings
Catalog No. 50-194-5115