Learn More
Sutter Instruments Thick/Standard Walled Borosilicate Glass with Filament, 225/pk
SDP
Supplier:  Sutter Instruments FGGBF1507510
Encompass_Preferred
Outer diameter1.50mm, Inner Diameter 0.75mm, Overall length 10cm, 225/pk
Catalog No. NC9544044