Learn More
Grainger Electrical Tape, Silicone Tape Adhesive, 7.0 mil Thick, 3/4 in X 66 ft, White
Supplier:  Grainger 2A458
Encompass_Preferred
Scotch™ 69 thermosetting silicone glass cloth tape features a thermosetting adhesive that provides an increased bond when exposed to high ambient temps. Use this electrical tape when you’re reinforcing insulation in electrical installations where heavy loads cause high heat and breakdown of the original insulation.
Catalog No. NC1076921