Learn More
Vee Gee Glass Erlenmeyer Flask, 500 mL
Supplier:  Vee Gee 10530500A/DEL
Encompass_Preferred
The Erlenmeyer Flask features heavy-duty rim to reduce chipping, white permanent graduations, and marking squares. It is made from high-quality borosilicate tubing with a low coefficient of expansion.• Uniform wall thickness - ideal for heating applications• Graduation accuracy is ±5% of full capacity • Rubber Stopper No.: 6 • Graduation Range: 200 to 500 mL • Graduation Intervals: 50 mL
Catalog No. NC1937426
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.