Learn More
Microsolv Technology Corporation Inserts, Low Volume for Vials, Glass. Clear, 300ul fill volume with a conical point and 6x38mm outer dimensions with attached plastic springs. For use in 4ml autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9504S04N
Encompass_Preferred
First hydrolytic, Class A, Type 33, Borosilicate Glass. Features a conical tip for excellent performance. Residual volume is 3ul with most autosamplers with a 200ul useable volume. Polypropylene springs are attached to keep the inserts center aligned and reduce chances of the needle breaking the inserts during use. This product is manufactured in an ISO 9001:2015 Certified facility. Use with 4ml, 13mm, Screw Top, 15x45mm autosampler vials.
Catalog No. 50-227-0546