Learn More
Vee Gee Vee Gee Glass Separatory Funnel, 2000 mL
Supplier:  Vee Gee 1426T2000/DEL
Encompass_Preferred
The Squibb Style Separatory Funnel with stopper features a smooth tooled tip, large white block letters, and large marking squares. Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination Made from high-quality borosilicate tubing with a low coefficient of expansion Stopper No.: 29/42 Stopcock Plug Size: 6
Catalog No. NC1937430