Learn More
Vee Gee Glass Separatory Funnel, 500 mL
Supplier:  Vee Gee 1426T500/DEL
Encompass_Preferred
The Squibb Style Separatory Funnel with stopper features a smooth tooled tip, large white block letters, and large marking squares.• Supplied with PTFE stopcocks to eliminate freezing and lubricant contamination• Made from high-quality borosilicate tubing with a low coefficient of expansion • Stopper No.: 24/40 • Stopcock Plug Size: 4
Catalog No. NC1937432