Learn More
Cole-Parmer Glass Tempering Beaker, borosilicate, 1/2" hose barbs, 600 mL
Supplier:  Cole-Parmer 0377320
Encompass_Preferred
Liquid-jacketed borosilicate glass vessels have double-walled casing for circulating cold or warm tempering fluid. Tubing connectors measure 1 1⁄2"L x 1⁄2" OD. Connect a constant temperature circulator to maintain sample temperatures.
Catalog No. NC9899332