Learn More
Essendant GLASSESANTI-FOG12/CSMK
Supplier:  Essendant ANS55391
Encompass_Preferred
KleenGuard Personal Protective Equipment (PPE) enables rather than inhibits allowing for maximum productivity and safety for you and your employees KleenGuard Nemesis Safety Glasses with Cord Connect allow for easy attatchment of earplug cords (not included) to the glasses to keep them untangled and ready for use The stylish design and durable lightweight construction are just some of the reasons why millions of people wear KleenGuard Nemesis Safety Glasses for protection both at work and at home These safety glasses have polycarbonate lenses that provide 99 9pct UVA/ UVB/UVC protection to help prevent eye damage like cataracts retinal damage and other conditions that can cause temporary vision loss The lenses are coated with KleenVision premium anti-fog coating that passes the EN166 N mark test for resistance to fogging The wraparound frame has a base curve of 8 that extends the range of peripheral protection as well as soft touch temples for comfort KleenGuard Nemesis Sa
Catalog No. 50-000-23067