Learn More
Essendant GLASSESELITE SMOKE W/AF
Supplier:  Essendant ANS21303
Encompass_Preferred
Lightweight retro-style safety glasses make a fashion statement while providing protection Wide temples improve stability and structural integrity Lenses are cut to provide clear distortion-free vision Dual lenses with 8-base curve and wraparound coverage High-performance polycarbonate lenses give 99 9pct UV protection Eyewear meets ANSI Z87 1plus impact standard Safety Glasses Style- Wraparound Frame Color- Black Lens Material- Polycarbonate Lens Tint- Smoke
Catalog No. 50-000-20411