Learn More
Essendant GLASSESHELLRSRSFTYSMK
Supplier:  Essendant ANS25714
Encompass_Preferred
Safety glasses feature a streamlined modern style for a sunglass-like appearance Extremely lightweight design makes them comfortable for extended wear Durable high-performance polycarbonate lenses Flex point temples increase comfort and stability Glasses meet ANSI Z87 1plus standard for impact resistance Safety Glasses Style- Wraparound Frame Color- Black Lens Material- Polycarbonate Lens Tint- Smoke
Catalog No. 50-000-22919