Learn More
Essendant GLASSESMAGNIFIER1.5 CLR
Supplier:  Essendant CRWBKH15
Encompass_Preferred
Well-thought-out construction combines the best in safety comfort and affordability Wraparound lenses won t obstruct vision fits snugly for maximum safety Comfort is enhanced by flexible temples and nonslip rubber head grips Filters 99 9pct of UV rays Magnifier Lens1 5 diopter strength Safety Glasses Style- Magnifier Frame Color- Clear Frame Material- Polycarbonate Lens Material- Polycarbonate
Catalog No. 50-000-18853