Learn More
Essendant GLASSESOTGSFTYBK/CLRAF
Supplier:  Essendant ANS20746
Encompass_Preferred
Stylish safety eyewear is designed to fit over most prescription eyeglasses Integral side shields offer increased protection Lenses offer 99pct UV protection Eyewear meets ANSI Z87 1plus and CSA Z94 3 standards Safety Glasses Style- Over the Glasses Frame Color- Black Frame Material- Nylon Lens Material- Polycarbonate
Catalog No. 50-000-22827